cis-4-Hydroxy-D(+)-proline 99%

Supplier: Thermo Scientific Chemicals

20491-0010 20491-0100
TS20491-0010EA 69 USD
TS20491-0010 TS20491-0100
cis-4-Hydroxy-D(+)-proline 99%
cis-4-Hydroxy-D(+)-proline
Formula: C₅H₉NO₃
Melting Pt: 243 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005252
CAS Number: 2584-71-6
EINECS: 219-963-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR