D(-)-Lysine monohydrochloride 99+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-(-)-Lysine monohydrochloride, H-D-Lys-OH.HCl, D-Lysine hydrochloride, (R)-(-)-2,6-Diaminocaproic acid monohydrochloride, (R)-(-)-2,6-Diaminohexanoic acid monohydrochloride
24308-0010
24308-0100
24308-0500
TS24308-0010EA
56
USD
TS24308-0010
TS24308-0100
TS24308-0500
D(-)-Lysine monohydrochloride 99+%
D(-)-Lysine monohydrochloride
Formula:
C₆H₁₅ClN₂O₂ MW: 182.65 g/mol Melting Pt: 266 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00012920 CAS Number: 7274-88-6 EINECS: 230-691-6 |
Specification Test Results
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (105°C, 3 h) |
Sulfated ash | ≤0.5 % |
Specific optical rotation | -19° to -22° (20°C, 589 nm) (c=5, 5 N HCl) |
Titration with HClO4 | 99.0 to 101.0 % |
Arsenic (As) | ≤1 ppm |
Iron (Fe) | ≤30 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...