L-Lysine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-2,6-Diaminohexanoic acid, L(+)-Lysine, (S)-(+)-2,6-Diaminohexanoic acid, (S)-(+)-Lysine, (S)-(+)-2,6-Diaminocaproic acid, H-Lys-OH
30334-1000
30334-5000
TS30334-1000EA
253
USD
TS30334-1000
TS30334-5000
L-Lysine 98%
L-Lysine
Formula:
C₆H₁₄N₂O₂ Melting Pt: 214…216 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064433 CAS Number: 56-87-1 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤5.0 % (K.F.) |
Specific optical rotation | +23° to +27° (20°C, 589 nm) (c=2, 1N HCl) |
Titration with HClO4 | ≥97.5 % (on dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...