DL-Proline 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-Proline, H-DL-Pro-OH

17403-0050 17403-0250
TS17403-0050EA 44 USD
TS17403-0050 TS17403-0250
DL-Proline 99%
DL-Proline
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 208…210 °C
MDL Number: MFCD00005250
CAS Number: 609-36-9
EINECS: 210-189-3
Merck Index: 12,07963

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.1 %
Solubility (5 % in water) Colorless to light yellow
Titration with HClO4 98.5 to 101.5 %
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR