DL-Lysine monohydrochloride 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(±)-2,6-Diaminocaproic acid monohydrochloride, H-DL-Lys-OH · HCl, H-DL-Lys-OH.HCl, (±)-2,6-Diaminohexanoic acid monohydrochloride, (±)-Lysine monohydrochloride
41337-0010
41337-0250
41337-1000
TS41337-0010EA
864
USD
TS41337-0010
TS41337-0250
TS41337-1000
DL-Lysine monohydrochloride 99%
DL-Lysine monohydrochloride
Formula:
C₆H₁₅ClN₂O₂ Melting Pt: 267 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064563 CAS Number: 70-53-1 EINECS: 200-739-0 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Specific optical rotation | -1° to +1° (20°C, 589 nm) (c=1, water) |
Titration with HClO4 | 98.5 to 101.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...