γ-(N-Morpholino)propanesulfonic acid (MOPS) 99.5% for molecular biology, for peptide sequencing DNase-, RNase-, protease-free
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
32766-5000
32766-1000
TS32766-5000EA
761
USD
TS32766-5000
TS32766-1000
γ-(N-Morpholino)propanesulfonic acid (MOPS) 99.5% for molecular biology, for peptide sequencing DNase-, RNase-, protease-free
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula:
C₇H₁₅NO₄S MW: 209.27 g/mol Melting Pt: 277…282 °C Flash Pt: 116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006183 CAS Number: 1132-61-2 EINECS: 214-478-5 Merck Index: 13,06288 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Assay | ≥99.5 % |
Loss on drying | ≤0.5 % (105°C) |
Solubility | (10 % in water) Complete |
UV | at 280 nm A: ≤0.01 |
pH | 3.0 to 4.5 (1 % at 25°C) |
Dnase,Rnase,Protease activity | none detected |
Heavy metals (as Pb) | ≤5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...