α-Linolenic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
30282-0050
30282-0010
30282-5000
TS30282-0050EA
503
USD
TS30282-0050
TS30282-0010
TS30282-5000
α-Linolenic acid 99%
α-Linolenic acid
Formula:
C₁₈H₃₀O₂ Boiling Pt: 230…232 °C (1 mmHg) Melting Pt: –11 °C Density: 0.91 g/cm³ Flash Pt: >112 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00065720 CAS Number: 463-40-1 EINECS: 207-334-8 Merck Index: 12,05530 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4790 to 1.4810 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...