2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid sodium salt 99%

Supplier: Thermo Scientific Chemicals

43903-0250 43903-1000 43903-5000
TS43903-0250EA 66 USD
TS43903-0250 TS43903-1000 TS43903-5000
2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid sodium salt 99%
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid sodium salt
Formula: C₆H₁₄NNaO₅S
Melting Pt: 155.6…156.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00067425
CAS Number: 66992-27-6

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Elemental Analysis 5.85 to 6.04 % (N)
Titration with HCl 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR