β-(N-Morpholino)ethanesufonic acid (MES) monohydrate 99+% for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate

32776-5000 32776-0025 32776-1000
TS32776-5000EA 670 USD
TS32776-5000 TS32776-0025 TS32776-1000
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate 99+% for biochemistry
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Formula: C₆H₁₃NO₄S·1H₂O
MW: 213.25 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water 8.0 to 9.0 % (K.F.)
UV at 280 nm A: ≤0.02
pH 2.5 to 4.0 (1 % in water at 25°C)
Titration with NaOH ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR