(S)-(-)-Nicotine 99+%

Supplier: Thermo Scientific Chemicals
Danger

18142-0250 18142-0050 18142-1000 18142-5000
TS18142-0250EA 218 USD
TS18142-0250 TS18142-0050 TS18142-1000 TS18142-5000
(S)-(-)-Nicotine 99+%
(S)-(-)-Nicotine
Formula: C₁₀H₁₄N₂
Boiling Pt: 247 °C (760 mmHg)
Melting Pt: –80 °C
Density: 1.015 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00006369
CAS Number: 54-11-5
EINECS: 200-193-3
UN: 1654
ADR: 6.1,II
Merck Index: 12,06611

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR