4-Ethoxybenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

43497-0010 43497-0050
TS43497-0010EA 52 USD
TS43497-0010 TS43497-0050
4-Ethoxybenzoyl chloride 98%
4-Ethoxybenzoyl chloride
Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 128…130 °C (6Torr)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000688
CAS Number: 16331-46-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR