D-Methionine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: D(+)-Methionine, (R)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-2-Amino-4-(methylthio)butyric acid, (R)-(+)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-Methionine, H-D-Met-OH

22721-0050 22721-0250 22721-1000
TS22721-0050EA 54 USD
TS22721-0050 TS22721-0250 TS22721-1000
D-Methionine 99+%
D-Methionine
Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Melting Pt: 273…275 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00002622
CAS Number: 348-67-4
EINECS: 206-483-6
Merck Index: 13,06004

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.3 %
Sulfated ash ≤0.1 %
UV transmittance ≥98 % (c=3, H2O, 430 nm) 1 cm cell
Specific optical rotation -23° to -25° (20°C, 589 nm) (c=8, 6 N HCl) (on dried substance)
Titration with HClO4 99.0 to 101.0 % (on dried substance)
Chloride (Cl) ≤300 ppm
Sulfate (SO4) ≤300 ppm
Ammonium (NH4) ≤200 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids (TLC) not detectable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR