N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES) 99%, extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid

22800-0250 22800-1000 22800-2500
TS22800-0250EA 53 USD
TS22800-0250 TS22800-1000 TS22800-2500
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES) 99%, extra pure
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES)
Formula: H₂NCOCH₂NHCH₂CH₂SO₃H
Melting Pt: 270…295 °C
Storage Temperature: Ambient
MDL Number: MFCD00008030
CAS Number: 7365-82-4
EINECS: 230-908-4
Merck Index: 13,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8d301fc0de89d24309fbfdbaedb924c1debb4a&vid=a5fb2045198b642d124372f31d901db2e2d25597&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
UV at 280 nm A: ≤0.02
pH 3.6 to 4.4 (1 % solution)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR