ADA 99% for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid

22801-0250 22801-1000
TS22801-0250EA 40 USD
TS22801-0250 TS22801-1000
ADA 99% for biochemistry
ADA
Formula: H₂NC(O)CH₂N(CH₂COOH)₂
MW: 190.16 g/mol
Melting Pt: 219 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008031
CAS Number: 26239-55-4
EINECS: 247-530-0
Merck Index: 13,00148

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (110°C, 5 hrs)
Sulfated ash ≤0.1 %
UV at 300 nm A: ≤0.08
Appearance of solution (9% soln. 1 M NaOH) clear, colorless
Titration with NaOH ≥98.5 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR