D-(+)-Tyrosine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-D-Tyr-OH

22691-0025 22691-0100
TS22691-0025EA 61 USD
TS22691-0025 TS22691-0100
D-(+)-Tyrosine 98%
D-(+)-Tyrosine
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351
Melting Pt: 285
Storage Temperature: Ambient
MDL Number: MFCD00063073
CAS Number: 556-02-5
EINECS: 209-112-6
Merck Index: 12,09970

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 3 h)
Solubility (5 % in 1 M HCl) clear colorless to almost colorless
Residue after ignition ≤0.1 %
Specific optical rotation +10° to +12° (20°C, 589 nm) (c=5, 1 M HCl, after 3 h)
Titration with HClO4 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR