L-Norvaline 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid

22602-0010 22602-0050 22602-0500
TS22602-0010EA 36 USD
TS22602-0010 TS22602-0050 TS22602-0500
L-Norvaline 99%
L-Norvaline
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064421
CAS Number: 6600-40-4
EINECS: 229-543-3
Merck Index: 12,06813

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless
Specific optical rotation +22.0° to +26.0° (20°C, 589 nm) (c=2, 5 N HCl)
Titration with HClO4 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR