L(+)-Valine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Val-OH, Valine, (S)-(+)-Valine

14081-0025 14081-5000 14081-0250 14081-1000
TS14081-0025EA 959 USD
TS14081-0025 TS14081-5000 TS14081-0250 TS14081-1000
L(+)-Valine 99%
L(+)-Valine
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 217
Melting Pt: 86
Density: 1.23 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6
Merck Index: 13,09458

Order Now

Specification Test Results

Appearance White Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 %
Sulfated ash ≤0.1 %
Specific optical rotation +26.5° to +29.0° (20°C, 589 nm) (c=8, 6 N HCl)
Titration with HClO4 ≥98.5 %
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤300 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR