D-(-)-Valine 98+%

Supplier: Thermo Scientific Chemicals

22722-0050 22722-0250
TS22722-0050EA 38 USD
TS22722-0050 TS22722-0250
D-(-)-Valine 98+%
D-(-)-Valine
Formula: C₅H₁₁NO₂
Melting Pt: 302…303 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064219
CAS Number: 640-68-6
EINECS: 211-368-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
Loss on drying ≤0.5 % (105°C, 4 hrs)
Sulfated ash ≤0.2 %
TLC ≤1 % L-enantiomer
Specific optical rotation -26° to -29° (20°C, 589 nm) (c=5, 5 N HCl)
Titration with HClO4 98.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR