DL-m-Tyrosine 98.5%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxyphenylalanine, 3-(m-Hidroxifenil)-DL-alanina, 3-(3-Hydroxyphenyl)-DL-alanine

14063-0010 14063-0050
TS14063-0010EA 77 USD
TS14063-0010 TS14063-0050
DL-m-Tyrosine 98.5%
DL-m-Tyrosine
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351
Melting Pt: 283 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002597
CAS Number: 775-06-4
EINECS: 212-270-9
Merck Index: 12,09971

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Titration with HClO4 ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR