L(-)-Xylose 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3R,4S)-2,3,4,5-Tetrahydroxypentanal, L-(-)-Xylose

22599-0050 22599-0250 22599-1000
TS22599-0050EA 53 USD
TS22599-0050 TS22599-0250 TS22599-1000
L(-)-Xylose 99+%
L(-)-Xylose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00151096
CAS Number: 609-06-3
EINECS: 210-174-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥99.0 % (ELSD)
Specific optical rotation -18° to -20° (20°C, 589 nm) (c=10, H2O) (after 24 h)
Loss on ignition ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR