L(-)-Mannose 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose

22759-0010 22759-0050 22759-2500
TS22759-0010EA 114 USD
TS22759-0010 TS22759-0050 TS22759-2500
L(-)-Mannose 99+%
L(-)-Mannose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00136021
CAS Number: 10030-80-5
EINECS: 233-080-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥99.0 % (ELSD)
Specific optical rotation -13.5° to -15.5° (20°C, 589 nm) (c=4, water, NH3)
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR