L(-)-Glucose 98%
Supplier: Thermo Scientific Chemicals
24192-0010
24192-0050
24192-0250
24192-2500
TS24192-0010EA
113
USD
TS24192-0010
TS24192-0050
TS24192-0250
TS24192-2500
L(-)-Glucose 98%
L(-)-Glucose
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 153…156 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00148913 CAS Number: 921-60-8 EINECS: 213-068-3 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Solubility | (10 % in water) Clear colorless |
HPLC | ≥97.5 % (ELSD) |
Specific optical rotation | -50° to -54° (20°C, 589 nm) (c=10, H2O, NH3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...