L(+)-Arabinose 99+%
Supplier: Thermo Scientific Chemicals
10498-0250
10498-1000
10498-5000
TS10498-0250EA
113
USD
TS10498-0250
TS10498-1000
TS10498-5000
L(+)-Arabinose 99+%
L(+)-Arabinose
Formula:
C₅H₁₀O₅ Melting Pt: 155…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067709 CAS Number: 5328-37-0 EINECS: 226-214-6 |
|
Specification Test Results
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % (K.F.) |
Sulfated ash | ≤0.3 % |
Solubility | (10 % in water) Clear colorless |
HPLC | ≥99.0 % (ELSD) |
Specific optical rotation | after 24 h |
Sodium (Na) | ≤0.2 % |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...