D-(+)-Maltose monohydrate 92+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
32991-0025
32991-1000
32991-5000
TS32991-0025EA
298
USD
TS32991-0025
TS32991-1000
TS32991-5000
D-(+)-Maltose monohydrate 92+%
D-(+)-Maltose monohydrate
Formula:
C₁₂H₂₂O₁₁·1H₂O MW: 360.32 g/mol Melting Pt: 130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149343 CAS Number: 6363-53-7 EINECS: 200-716-5 Merck Index: 13,05736 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤7 % |
Sulfated ash | ≤0.1 % |
Additional info | remainder mainly glucose and maltotriose |
HPLC | ≥92.0 % |
Specific optical rotation | +135° to +139° (20°C, 589 nm) (c=4, H2O, NH3) (on dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...