Linoleic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid

21504-1000 21504-0050 21504-0250
TS21504-1000EA 394 USD
TS21504-1000 TS21504-0050 TS21504-0250
Linoleic acid 99%
Linoleic acid
Formula: C₁₈H₃₂O₂
Boiling Pt: 229…230 °C (16 mmHg)
Melting Pt: –5 °C
Density: 0.9 g/cm³
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064241
CAS Number: 60-33-3
EINECS: 200-470-9
Merck Index: 13,05527

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4687 to 1.4707 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR