3-Isobutyl-1-methylxanthine 99+%

Supplier: Thermo Scientific Chemicals
Warning

22842-0010 22842-0050
TS22842-0010EA 214 USD
TS22842-0010 TS22842-0050
3-Isobutyl-1-methylxanthine 99+%
3-Isobutyl-1-methylxanthine
Formula: C₁₀H₁₄N₄O₂
Melting Pt: 200…203 °C
Storage Temperature: Freezer
MDL Number: MFCD00005584
CAS Number: 28822-58-4
EINECS: 249-259-3

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 200 to 203 °C
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR