L(+)-Norleucine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Nle-OH, H-NorLeu-OH, (S)-2-Aminohexanoic acid, (S)-(+)-Norleucine

22604-0010 22604-0050
TS22604-0010EA 53 USD
TS22604-0010 TS22604-0050
L(+)-Norleucine 99+%
L(+)-Norleucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064423
CAS Number: 327-57-1
EINECS: 206-321-4
Merck Index: 13,06738

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C, 4 hrs)
Specific optical rotation +22° to +24° (20°C, 589 nm) (c=2, 6 N HCl)
Titration with HClO4 ≥99.0 %
Other amino acids (TLC) Not detectable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR