D-(-)-Leucine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-D-Leu-OH, (R)-(-)-Leucine

22690-0025 22690-0100
TS22690-0025EA 47 USD
TS22690-0025 TS22690-0100
D-(-)-Leucine 99%
D-(-)-Leucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 226
Melting Pt: >300 °C
MDL Number: MFCD00063088
CAS Number: 328-38-1
EINECS: 206-327-7

Order Now

Specification Test Results

Appearance White Powder or crystals or flakes
Infrared spectrum Conforms
Loss on drying ≤0.2 %
TLC ≤0.5 % L-enantiomer
Appearance of solution (5% soln. in 3 N HCl) clear, colorless
Residue after ignition ≤0.1 %
Specific optical rotation -14.9° to -16° (20°C, 589 nm) (c=4, 6 N HCl)
Titration with HClO4 ≥98.5 %
Arsenic (As) ≤2 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR