L(+)-allo-Isoleucine 99%, 99% ee

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3R)-(+)-allo-Isoleucine, L-Alloisoleucine, (2S,3R)-2-Amino-3-methylpentanoic acid

39949-0010 39949-1000
TS39949-0010EA 787 USD
TS39949-0010 TS39949-1000
L(+)-allo-Isoleucine 99%, 99% ee
L(+)-allo-Isoleucine
Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17 g/mol
Melting Pt: 285 °C
Storage Temperature: Freezer
MDL Number: MFCD00066446
CAS Number: 1509-34-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
NMR consistent with structure
Enantiomeric excess ≥98.5 % (HPLC)
Specific optical rotation +35° to +39° (20°C, 589 nm) (C=5, 1N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR