D(-)-allo-Isoleucine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-D-Allo-Ile-OH, (2R,3S)-(-)-allo-Isoleucine, H-D-allo-Ile-OH

35743-0010 35743-5000
TS35743-0010EA 280 USD
TS35743-0010 TS35743-5000
D(-)-allo-Isoleucine 97%
D(-)-allo-Isoleucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00066445
CAS Number: 1509-35-9
EINECS: 216-143-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Specific optical rotation -35° to -39° (20°C, 589 nm) (C=5, 1 N HCl)
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR