3-Methyladenine 90+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methyl-3H-purin-6-amine, 3-Methyl-3H-adenine

37979-1000 37979-5000
TS37979-1000EA 166 USD
TS37979-1000 TS37979-5000
3-Methyladenine 90+%
3-Methyladenine
Formula: C₆H₇N₅
MW: 149.16 g/mol
Melting Pt: 300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010531
CAS Number: 5142-23-4
EINECS: 225-908-6

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR