L(+)-Asparagine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: L-Asparagine, (S)-(+)-Asparagine, H-Asn-OH

37160-0025 37160-1000 37160-5000
TS37160-0025EA 915 USD
TS37160-0025 TS37160-1000 TS37160-5000
L(+)-Asparagine 99%
L(+)-Asparagine
Formula: H₂NCOCH₂CH(NH₂)CO₂H
Storage Temperature: Ambient
MDL Number: MFCD00064401
CAS Number: 70-47-3
EINECS: 200-735-9
Merck Index: 12,00872
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&vid=3d09743d0834d64e4719e8295f634c39195c571f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
Specific optical rotation +33° to +36° (20°C, 589 nm) (c=10, 2N HCl)
Titration with HClO4 ≥98.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR