D-(+)-Raffinose pentahydrate 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: D-(+)-Raffinose pentahydrate, Gossypose pentahydrate, (3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrate, Melitriose pentahydrate, O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose pentahydrate

19567-1000 19567-5000 19567-0250
TS19567-1000EA 276 USD
TS19567-1000 TS19567-5000 TS19567-0250
D-(+)-Raffinose pentahydrate 99+%
D-(+)-Raffinose pentahydrate
Formula: C₁₈H₃₂O₁₆·5H₂O
MW: 594.52 g/mol
Melting Pt: 78…80.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00071590
CAS Number: 17629-30-0
EINECS: 208-146-9
Merck Index: 12,08279

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water 14 to 16 % (K.F.)
Sulfated ash ≤0.1 % (on dry basis)
Solubility (10 % in water) Clear colorless
HPLC ≥99.0 % (ELSD)
Specific optical rotation +103.0° to +107.0° (20°C, 589 nm) (c=10, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR