Agar, Bacteriological Grade
Supplier: Thermo Scientific Chemicals
Synonyms:
Agar Agar
44357-0050
44357-0010
44357-2500
TS44357-0050EA
1295
USD
TS44357-0050
TS44357-0010
TS44357-2500
Agar, Bacteriological Grade
Agar
Storage Temperature:
Ambient |
MDL Number:
MFCD00081288 CAS Number: 9002-18-0 |
Specification Test Results
Appearance | White to cream to light yellow to beige Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤15 % (1 g, 105°C) |
Solubility | (1.5 % in hot water) Clear to slightly opalescent light yellow to yellow solution |
Gel strength | ≥700 g/cm² (1.5 %) |
Gel point | 32 to 38°C (1.5 %) |
Ash | ≤5 % |
pH | 6.0 to 7.5 (10 % suspension) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...