1-Methyladenine 98+%

Supplier: Thermo Scientific Chemicals
Warning

20131-0250 20131-1000 20131-2500
TS20131-0250EA 134 USD
TS20131-0250 TS20131-1000 TS20131-2500
1-Methyladenine 98+%
1-Methyladenine
Formula: C₆H₇N₅
MW: 149.16 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00010532
CAS Number: 5142-22-3
EINECS: 225-907-0

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR