Disulfiram 97%

Supplier: Thermo Scientific Chemicals
Warning

13812-0025 13812-0250 13812-5000
TS13812-0025EA 338 USD
TS13812-0025 TS13812-0250 TS13812-5000
Disulfiram 97%
Disulphiram
Formula: C₁₀H₂₀N₂S₄
Boiling Pt: 117 °C (23hPa)
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00009048
CAS Number: 97-77-8
EINECS: 202-607-8
UN: 3077
ADR: 9,III
Merck Index: 13,03399

Order Now

Specification Test Results

Appearance White to cream to light yellow Crystalline powder or crystals or granules
Infrared spectrum Conforms
Melting point 67 to 74 °C
Water ≤1.0 % (KF)
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR