D-(-)-Aspartic acid 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH

20426-0050 20426-0250 20426-1000
TS20426-0050EA 34 USD
TS20426-0050 TS20426-0250 TS20426-1000
D-(-)-Aspartic acid 99+%
D-(-)-Aspartic acid
Formula: C₄H₇NO₄
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00063081
CAS Number: 1783-96-6
EINECS: 217-234-6
Merck Index: 13,00845

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.4 % (105°C, 3 hrs)
Enantiomeric excess ≤0.5 % (L-enantiomer)
Residue after ignition ≤0.1 %
Specific optical rotation -23.8° to -25.8° (20°C, 589 nm) (c=5, 5 N HCl)
Titration with NaOH ≥99.0 %
Other amino acids ≤0.3 % (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR