DL-Tryptophan 98%

Supplier: Thermo Scientific Chemicals

17211-0250 17211-1000
TS17211-0250EA 76 USD
TS17211-0250 TS17211-1000
DL-Tryptophan 98%
DL-Tryptophan
Formula: C₁₁H₁₂N₂O₂
Melting Pt: 289…290 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064339
CAS Number: 54-12-6
EINECS: 200-194-9

Order Now

Specification Test Results

Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.8 % (105°C, 3 h) (Vacuum)
Solubility (1 % in 1 M HCl) Clear to slightly hazy colorles to yellow
Titration with HClO4 ≥97.5 %
Other amino acids (TLC) Not detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR