D-(-)-Glutamine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-D-Gln-OH, (R)-2,5-Diamino-5-oxopentanoic acid, (R)-(-)-Glutamine

13516-0010 13516-0050
TS13516-0010EA 77 USD
TS13516-0010 TS13516-0050
D-(-)-Glutamine 98%
D-(-)-Glutamine
Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
MDL Number: MFCD00065607
CAS Number: 5959-95-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Specific optical rotation -30° to -34° (20°C, 589 nm) (c=10, 2 N HCl)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR