5-Bromo-DL-tryptophan 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Amino-3-(5-bromo-1H-indol-3-yl)propanoic acid

10751-0010 10751-0050 10751-5000
TS10751-0010EA 296 USD
TS10751-0010 TS10751-0050 TS10751-5000
5-Bromo-DL-tryptophan 99%
5-Bromo-DL-tryptophan
Formula: C₁₁H₁₁BrN₂O₂
MW: 283.12 g/mol
Melting Pt: 261…267 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00005648
CAS Number: 6548-09-0
EINECS: 229-464-4

Order Now

Specification Test Results

Appearance White to light beige to light grey Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Titration with HClO4 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR