D-(+)-Sucrose octaacetate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
D-(+)-Saccharose octaacetate, Sucrose octaacetate, D(+)-Sucroseoctaacetat, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-(((2S,3S,4R,5R)-3,4-diacetoxy-2,5-bis(acetoxymethyl)tetrahydrofuran-2-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate
41979-0010
41979-0051
41979-2500
TS41979-0010EA
107
USD
TS41979-0010
TS41979-0051
TS41979-2500
D-(+)-Sucrose octaacetate 98%
D(+)-Sucrose octaacetate
Formula:
C₂₈H₃₈O₁₉ MW: 678.6 g/mol Boiling Pt: 250 °C (1 mmHg) Melting Pt: >78 °C Density: 1.28 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00006623 CAS Number: 126-14-7 EINECS: 204-772-1 Merck Index: 12,09052 |
Specification Test Results
Appearance | White to cream Powder |
Infrared spectrum | Conforms |
Melting point | ≥78 °C |
Water | ≤1 % (K.F.) |
Residue after ignition | ≤0.1 % |
Titration excess NaOH | 97.5 to 102.5 % (on anhydrous substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...