Undecanoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
17397-0050
17397-0250
17397-1000
TS17397-0050EA
26
USD
TS17397-0050
TS17397-0250
TS17397-1000
Undecanoic acid 99%
Undecanoic acid
Formula:
C₁₁H₂₂O₂ Boiling Pt: 228 °C (1013 hPa) Melting Pt: 28.5 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002730 CAS Number: 112-37-8 EINECS: 203-964-2 |
Specification Test Results
Appearance | White to light yellow Low melting solid |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...