Myristic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetradecylic acid, Tetradecanoic acid

15696-0010 15696-1000 15696-2500
TS15696-0010EA 82 USD
TS15696-0010 TS15696-1000 TS15696-2500
Myristic acid 99%
Myristic acid
Formula: C₁₄H₂₈O₂
MW: 228.38 g/mol
Boiling Pt: 326 °C (760 mmHg)
Melting Pt: 50…57 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002744
CAS Number: 544-63-8
EINECS: 208-875-2
Merck Index: 12,06416

Order Now

Specification Test Results

Appearance White Powder or flakes or chunks or crystalline mass
Infrared spectrum Conforms
Melting point 50 to 57 °C
GC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR