Valeric acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pentanoic acid

14957-0010 14957-0025 14957-0050 14957-1000
TS14957-0010EA 88 USD
TS14957-0010 TS14957-0025 TS14957-0050 TS14957-1000
Valeric acid 99%
Valeric acid
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 185 °C (760 mmHg)
Melting Pt: 20…18 °C
Density: 0.94 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00004413
CAS Number: 109-52-4
EINECS: 203-677-2
UN: 3265
ADR: 8,II
Merck Index: 12,10042

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4100 (20°C, 589 nm)
Water ≤0.1 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR