Pentadecanoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

12983-0050 12983-0250
TS12983-0050EA 48 USD
TS12983-0050 TS12983-0250
Pentadecanoic acid 99%
Pentadecanoic acid
Formula: C₁₅H₃₀O₂
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 51…53 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002745
CAS Number: 1002-84-2
EINECS: 213-693-1

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
GC ≥98.5 %
Titration with NaOH ≥98.5 %
Specific optical rotation -63° to -67° (20°C, 589 nm) (c=4, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR