Caprylic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid

12939-0010 12939-0025 12939-0251
TS12939-0010EA 58 USD
TS12939-0010 TS12939-0025 TS12939-0251
Caprylic acid 99%
Caprylic acid
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 238 °C (760 mmHg)
Melting Pt: 16…16.5 °C
Density: 0.91 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8,III
Merck Index: 13,01771

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4300 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR