Nonadecanoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

20719-0010 20719-0050
TS20719-0010EA 59 USD
TS20719-0010 TS20719-0050
Nonadecanoic acid 98+%
Nonadecanoic acid
Formula: C₁₉H₃₈O₂
Boiling Pt: 297 °C (100 mmHg)
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00002754
CAS Number: 646-30-0
EINECS: 211-472-4

Order Now

Specification Test Results

Appearance White Glistening powder or flakes
Infrared spectrum Conforms
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR