Caproic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexanoic acid

12070-0010 12070-0025 12070-2500 12070-5000
TS12070-0010EA 112 USD
TS12070-0010 TS12070-0025 TS12070-2500 TS12070-5000
Caproic acid 99%
Caproic acid
Formula: C₆H₁₂O₂
Boiling Pt: 202…203 °C (760 mmHg)
Melting Pt: –3 °C
Density: 0.92 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00004421
CAS Number: 142-62-1
EINECS: 205-550-7
UN: 2829
ADR: 8,III
Merck Index: 13,01765

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4180 (20°C, 589 nm)
GC ≥98.5 %
Acid value 473 to 493 mg KOH/g
test 50 Result 50
test 51 Result 51
test 52 Result 52
test 53 Result 53
test 54 Result 54
test 55 Result 55
test 56 Result 56
test 57 Result 57


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR