Heptanoic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Heptylic acid, Enanthic acid

16417-0010 16417-0025 16417-2500
TS16417-0010EA 81 USD
TS16417-0010 TS16417-0025 TS16417-2500
Heptanoic acid 98%
Heptanoic acid
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 223…223.4 °C (760 mmHg)
Melting Pt: –10.5 °C
Density: 0.91 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004426
CAS Number: 111-14-8
EINECS: 203-838-7
UN: 3265
ADR: 8,III
Merck Index: 13,04680

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4210 to 1.4250 (20°C, 589 nm)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR