Heneicosanoic acid 99%

Supplier: Thermo Scientific Chemicals

20469-0010 20469-0050 20469-1000
TS20469-0010EA 157 USD
TS20469-0010 TS20469-0050 TS20469-1000
Heneicosanoic acid 99%
Heneicosanoic acid
Formula: C₂₁H₄₂O₂
Boiling Pt: 170 °C (0.1 mbar)
Melting Pt: 74…76 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002805
CAS Number: 2363-71-5
EINECS: 219-113-3

Order Now

Specification Test Results

Appearance White Glistening crystalline powder
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR